missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-1-butanol 98.0+%, TCI America™

Catalog No. D16595ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16595ML 5 mL
D165925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16595ML Supplier TCI America Supplier No. D16595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13330-96-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00020624
InChI Key QCTOLMMTYSGTDA-UHFFFAOYSA-N
PubChem CID 83350
IUPAC Name 4-(dimethylamino)butan-1-ol
SMILES CN(C)CCCCO

Specifications

Color Colorless
Boiling Point 188°C
Quantity 5 mL
UN Number 2735
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 4-Dimethylamino-1-butanol