accessibility menu, dialog, popup

UserName

CAS RN 132765-35-6

IUPAC Name: 3-({5-[(2-cyanoethyl)sulfanyl]-2-sulfanylidene-2H-1,3-dithiol-4-yl}sulfanyl)propanenitrile
Synonyms: 3-({5-[(2-cyanoethyl)sulfanyl]-2-sulfanylidene-1,3-dithiol-4-yl}sulfanyl)propanenitrile
Molecular Weight (g/mol): 304.48
Molecular Formula: C9H8N2S5
InChi Key: DRMGXZWMQXEMKB-UHFFFAOYSA-N
SMILES: S=C1SC(SCCC#N)=C(SCCC#N)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results