missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(2-cyanoethylthio)-1,3-dithiole-2-thione 98.0+%, TCI America™

Catalog No. B22281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22281G Supplier TCI America Supplier No. B22281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132765-35-6
Molecular Formula C9H8N2S5
Molecular Weight (g/mol) 304.477
MDL Number MFCD01096160
InChI Key DRMGXZWMQXEMKB-UHFFFAOYSA-N
PubChem CID 2818208
IUPAC Name 3-[[5-(2-cyanoethylsulfanyl)-2-sulfanylidene-1,3-dithiol-4-yl]sulfanyl]propanenitrile
SMILES C(CSC1=C(SC(=S)S1)SCCC#N)C#N

Specifications

Melting Point 83°C
Quantity 1 g
UN Number 2811
Formula Weight 304.48
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Lumps
Chemical Name or Material 4,5-Bis(2-cyanoethylthio)-1,3-dithiole-2-thione