accessibility menu, dialog, popup

UserName

CAS RN 132388-59-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)carbamoyl]propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(triphenylmethylcarbamoyl)propanoate
Molecular Weight (g/mol): 595.68
Molecular Formula: C38H31N2O5
InChi Key: KJYAFJQCGPUXJY-UMSFTDKQSA-M
SMILES: [O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)