accessibility menu, dialog, popup

UserName

CAS RN 13195-50-1

IUPAC Name: 2-bromo-5-nitrothiophene
Synonyms: 2-bromo-5-nitrothiophene
Molecular Weight (g/mol): 208.03
Molecular Formula: C4H2BrNO2S
InChi Key: ZPNFMDYBAQDFDY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)S1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)