missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiophene, 97%

Catalog No. AAA1586614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1586614 25 g
AAA1586606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1586614 Supplier Thermo Scientific Chemicals Supplier No. A1586614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13195-50-1 | C4H2BrNO2S | 208.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13195-50-1
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.029
MDL Number MFCD00022493
InChI Key ZPNFMDYBAQDFDY-UHFFFAOYSA-N
PubChem CID 83222
IUPAC Name 2-bromo-5-nitrothiophene
SMILES C1=C(SC(=C1)Br)[N+](=O)[O-]

Specifications

Melting Point 42°C to 46°C
Boiling Point 235°C to 237°C
Quantity 25 g
Beilstein 120955
Synonym 5-bromo-2-nitrothiophene, thiophene, 2-bromo-5-nitro, pubchem5283, acmc-1bsij, 2-nitro-5-bromothiophene, 5-nitro-2-bromothiophene, 5-bromo-2-nitro-thiophene, 2-bromo-5-nitro-thiophene, labotest-bb lt00455100, thiophene,2-bromo-5-nitro
Formula Weight 208.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-nitrothiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-155-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only