accessibility menu, dialog, popup

UserName

CAS RN 13171-00-1

CH3OH3CCH3CH3H3CCH3

CAS RN 13171-00-1

IUPAC Name: 1-(6-tert-butyl-1,1-dimethyl-2,3-dihydro-1H-inden-4-yl)ethan-1-one
Synonyms: celestolide
Molecular Weight (g/mol): 244.38
Molecular Formula: C17H24O
InChi Key: IKTHMQYJOWTSJO-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC2=C1CCC2(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results