missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-6-tert-butyl-1,1-dimethylindan 98.0+%, TCI America™

Numéro de catalogue. A220925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A220925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A220925G Fournisseur TCI America Code fournisseur A220925G
Il en reste null

Identifiants chimiques

CAS 13171-00-1
Molecular Formula C17H24O
Molecular Weight (g/mol) 244.38
MDL Number MFCD00046324
InChI Key IKTHMQYJOWTSJO-UHFFFAOYSA-N
PubChem CID 61585
IUPAC Name 1-(6-tert-butyl-1,1-dimethyl-2,3-dihydro-1H-inden-4-yl)ethan-1-one
SMILES CC(=O)C1=CC(=CC2=C1CCC2(C)C)C(C)(C)C

Spécifications

Melting Point 78°C
Color White-Yellow
Boiling Point 117°C
Quantity 25 g
Formula Weight 244.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetyl-6-tert-butyl-1,1-dimethylindan
EINECSNumber (4)-1202
TSCA Yes