accessibility menu, dialog, popup

UserName

CAS RN 13170-43-9

IUPAC Name: [(chloromagnesio)methyl]trimethylsilane
Synonyms: [(chloromagnesio)methyl]trimethylsilane
Molecular Weight (g/mol): 146.97
Molecular Formula: C4H11ClMgSi
InChi Key: NAQATMJWCJCHOZ-UHFFFAOYSA-M
SMILES: C[Si](C)(C)C[Mg]Cl
Quantity 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results