missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylmethylmagnesium Chloride (20% in Ethyl Ether, ca. 1mol/L), TCI America™

Catalog No. T1451100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1451100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1451100ML Supplier TCI America Supplier No. T1451100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13170-43-9
Molecular Formula C4H11ClMgSi
Molecular Weight (g/mol) 146.97
MDL Number MFCD00009916
InChI Key NAQATMJWCJCHOZ-UHFFFAOYSA-M
Synonym trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
PubChem CID 2734900
IUPAC Name [(chloromagnesio)methyl]trimethylsilane
SMILES C[Si](C)(C)C[Mg]Cl

Specifications

Quantity 100 mL
UN Number 3399
Formula Weight 146.97
Chemical Name or Material Trimethylsilylmethylmagnesium Chloride (20% in Ethyl Ether, ca. 1mol/L)