accessibility menu, dialog, popup

UserName

H3CONHOOHOHOHOHOOOONOONHOCH3OOOHOHOHOHOHHO

CAS RN 1316822-90-8

IUPAC Name: 5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 795.7
Molecular Formula: C31H45N3O21
InChi Key: OQZSJFGKSKBLDR-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)CC(OCC2OC(OC3=CC=C(C=C3)[N+]([O-])=O)C(NC(C)=O)C(OC3OC(CO)C(O)C(O)C3O)C2O)(OC1C(O)C(O)CO)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results