missing translation for 'onlineSavingsMsg'
Learn More

Gal beta(1-3)[Neu5Ac alpha(2-6)]GalNAc-alpha-pNP 95.0+%, TCI America™

Catalog No. G03772MG
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03772MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03772MG Supplier TCI America Supplier No. G03772MG

Chemical Identifiers

CAS 1316822-90-8
Molecular Formula C31H45N3O21
Molecular Weight (g/mol) 795.70
MDL Number MFCD20265590
InChI Key OQZSJFGKSKBLDR-UHFFFAOYNA-N
PubChem CID 133554216
IUPAC Name 5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
SMILES CC(=O)NC1C(O)CC(OCC2OC(OC3=CC=C(C=C3)[N+]([O-])=O)C(NC(C)=O)C(OC3OC(CO)C(O)C(O)C3O)C2O)(OC1C(O)C(O)CO)C(O)=O

Specifications

Color White-Yellow
Quantity 2 mg
Formula Weight 795.70
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-3)[Neu5Ac alpha(2-6)]GalNAc-alpha-pNP
TSCA No
Recommended Storage Freezer