accessibility menu, dialog, popup

UserName

CAS RN 13147-25-6

IUPAC Name: 2-[ethyl(nitroso)amino]ethan-1-ol
Synonyms: 2-[ethyl(nitroso)amino]ethanol
Molecular Weight (g/mol): 118.14
Molecular Formula: C4H10N2O2
InChi Key: HNQBPUIXFDQDRJ-UHFFFAOYSA-N
SMILES: CCN(CCO)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results