missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(2-hydroxyethyl)nitrosamine 97.0+%, TCI America™

Catalog No. E05805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05805G 5 g
E05801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05805G Supplier TCI America Supplier No. E05805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13147-25-6
Molecular Formula C4H10N2O2
Molecular Weight (g/mol) 118.136
MDL Number MFCD01727871
InChI Key HNQBPUIXFDQDRJ-UHFFFAOYSA-N
Synonym N-Ethyl-N-nitroso-2-aminoethanol
PubChem CID 25742
IUPAC Name N-ethyl-N-(2-hydroxyethyl)nitrous amide
SMILES CCN(CCO)N=O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 118.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-(2-hydroxyethyl)nitrosamine
RTECSNumber KL0350000
TSCA No
Recommended Storage Refrigerator