CAS RN 1313876-00-4
IUPAC Name:
4,7-dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole
Synonyms:
4,7-dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole
Molecular Weight (g/mol):
662.61
Molecular Formula:
C30H50Br2N2O2S
InChi Key:
DVDXEQCMUYXONV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCOC1=C(Br)C2=NSN=C2C(Br)=C1OCCCCCCCCCCCC
1
–
1
of
1
results
1
4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole 98.0+%, TCI America™
CAS: 1313876-00-4 Molecular Formula: C30H50Br2N2O2S Molecular Weight (g/mol): 662.61 InChI Key: DVDXEQCMUYXONV-UHFFFAOYSA-N PubChem CID: 71143603 IUPAC Name: 4,7-dibromo-5,6-didodecoxy-2,1,3-benzothiadiazole SMILES: CCCCCCCCCCCCOC1=C(C2=NSN=C2C(=C1OCCCCCCCCCCCC)Br)Br