missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. D4622200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4622200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4622200MG Supplier TCI America Supplier No. D4622200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1313876-00-4
Molecular Formula C30H50Br2N2O2S
Molecular Weight (g/mol) 662.61
InChI Key DVDXEQCMUYXONV-UHFFFAOYSA-N
PubChem CID 71143603
IUPAC Name 4,7-dibromo-5,6-didodecoxy-2,1,3-benzothiadiazole
SMILES CCCCCCCCCCCCOC1=C(C2=NSN=C2C(=C1OCCCCCCCCCCCC)Br)Br

Specifications

Melting Point 61°C
Color White
Quantity 200 mg
Formula Weight 662.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole