accessibility menu, dialog, popup

UserName

CAS RN 13129-17-4

IUPAC Name: 4,5,6,7-tetrahydro-1-benzothiophene
Synonyms: 4,5,6,7-tetrahydro-1-benzothiophene
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: CBKDCOKSXCTDAA-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)SC=C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results