missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene, 97%

Catalog No. AAH3430906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3430906 5 g
AAH3430903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3430906 Supplier Thermo Scientific Chemicals Supplier No. H3430906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13129-17-4 | C8H10S | 138.228 g/mol

It is an active pharmaceutical intermediate. Condensation of amino carboxyalkyl derivative of 4,5,6,7-tetrahydrobenzo[b]thiophene with salicylaldehyde yielded a potentially tridentate Schiff base (HSAT), which formed a series of copper(II) complexes. Reaction of ethyl 2-diazo-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 1 with 3-iminobutyronitrile 2 gave the hydrazone derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate. Condensation of amino carboxyalkyl derivative of 4,5,6,7-tetrahydrobenzo[b]thiophene with salicylaldehyde yielded a potentially tridentate Schiff base (HSAT), which formed a series of copper(II) complexes. Reaction of ethyl 2-diazo-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 1 with 3-iminobutyronitrile 2 gave the hydrazone derivative.

Notes
Air sensitive. Store at room temperature. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 13129-17-4
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD19442759
InChI Key CBKDCOKSXCTDAA-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene, benzothiophene, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-2-benzothiophene, benzo b thiophene, 4,5,6,7-tetrahydro, 5-17-01-00498 beilstein handbook reference
PubChem CID 22946
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene
SMILES C1CCC2=C(C1)C=CS2

Specifications

Boiling Point 55°C (1 mmHg)
Refractive Index 1.558
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only