accessibility menu, dialog, popup

UserName

CAS RN 13116-27-3

HNNH2I

CAS RN 13116-27-3

IUPAC Name: (4-iodophenyl)hydrazine
Synonyms: (4-iodophenyl)hydrazine
Molecular Weight (g/mol): 234.04
Molecular Formula: C6H7IN2
InChi Key: IQMLCMKMSBMMGR-UHFFFAOYSA-N
SMILES: NNC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results