accessibility menu, dialog, popup

UserName

CH3OOOOHOCH3OOCH3OOCH3O

CAS RN 13100-46-4

IUPAC Name: 4,5,6-tris(acetyloxy)-2-(hydroxymethyl)oxan-3-yl acetate
Synonyms: 4,5,6-tris(acetyloxy)-2-(hydroxymethyl)oxan-3-yl acetate
Molecular Weight (g/mol): 348.3
Molecular Formula: C14H20O10
InChi Key: FEQXFAYSNRWXDW-UHFFFAOYNA-N
SMILES: CC(=O)OC1OC(CO)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results