Learn More
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Description
Germanium dioxide is used as an optical material for wide-angle lenses and in optical microscope objective lenses. It is used as a catalyst in the preparation of polyethylene terephthalate resin. Further, it is used as a feedstock for the manufacturing of phosphors and semiconductor materials. It is also used in transistors and diodes, and infrared transmitting glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1086°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Odor | Odorless |
| Assay Percent Range | 99.999% (Metals basis) |
| Density | 6.239 |
| Solubility Information | Slightly soluble in sodium hydroxide. Insoluble in water and hydrochloric acid.Soluble in water,acid and alkali. Insoluble in hydrofluoric acid and hydrochloric acid. |
| Formula Weight | 104.59 |
| Chemical Name or Material | Germanium(IV) oxide |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| EINECSNumber | 215-180-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only