accessibility menu, dialog, popup

UserName

H2COOOOCH2

CAS RN 131-17-9

IUPAC Name: 1,2-bis(prop-2-en-1-yl) benzene-1,2-dicarboxylate
Synonyms: diallylphthalate
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: QUDWYFHPNIMBFC-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)