missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Phthalate 98.0+%, TCI America™

Catalog No. P0290500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0290500ML 500 mL
P029025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0290500ML Supplier TCI America Supplier No. P0290500ML

Chemical Identifiers

CAS 131-17-9
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00008646
InChI Key QUDWYFHPNIMBFC-UHFFFAOYSA-N
Synonym diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
PubChem CID 8560
IUPAC Name bis(prop-2-enyl) benzene-1,2-dicarboxylate
SMILES C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C

Specifications

Color Yellow
Boiling Point 158°C
Quantity 500 mL
UN Number 3082
Formula Weight 246.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Phthalate
EINECSNumber (3)-1325
RTECSNumber CZ4200000
TSCA Yes