Learn More
Identifiants chimiques
| CAS | 131-17-9 |
|---|---|
| Molecular Formula | C14H14O4 |
| Molecular Weight (g/mol) | 246.26 |
| MDL Number | MFCD00008646 |
| InChI Key | QUDWYFHPNIMBFC-UHFFFAOYSA-N |
| Synonym | diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st |
| PubChem CID | 8560 |
| IUPAC Name | bis(prop-2-enyl) benzene-1,2-dicarboxylate |
| SMILES | C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|
AC276622500
|
N/A | 250mL | Glass bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
AC276620010
|
N/A | 1L | Glass bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
AC276620025
|
N/A | 2.5L | Glass bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C14H14O4 | |
| MFCD00008646 | |
| diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
| bis(prop-2-enyl) benzene-1,2-dicarboxylate |
Spécifications
| 131-17-9 | |
| 300.0°C | |
| Authentic | |
| C14H14O4 | |
| MFCD00008646 | |
| 09, 4120 | |
| diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
| QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
| bis(prop-2-enyl) benzene-1,2-dicarboxylate | |
| 8560 | |
| 246.26 | |
| Diallyl phthalate |
| 1.1200g/mL | |
| 166°C | |
| 97.5% min. (GC) | |
| 1.5180 to 1.5200 | |
| 3082 | |
| 1.12 | |
| Solubility in water: 148mg/l (20°C) | |
| C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C | |
| 246.26 | |
| 13 mPa.s (20°C) | |
| 98% |
Sécurité et manipulation
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause damage to organs thr
GHS P Statement
Avoid release to the environment.
Do not get in eyes,on skin,or on clothing.
Call a POISON CENTRE or doctor/physician.
GHS Signal Word: Warning
EINECSNumber : 205-016-3
RUO – Research Use Only