missing translation for 'onlineSavingsMsg'
Learn More

Diallyl phthalate, 97%

Catalog No. AAB2464836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2464836 500 g
AAB2464822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2464836 Supplier Thermo Scientific Chemicals Supplier No. B2464836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 131-17-9 | C14H14O4 | 246.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131-17-9
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00008646
InChI Key QUDWYFHPNIMBFC-UHFFFAOYSA-N
Synonym diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
PubChem CID 8560
IUPAC Name bis(prop-2-enyl) benzene-1,2-dicarboxylate
SMILES C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C

Specifications

Melting Point -70°C
Density 1.12
Boiling Point 160°C to 163°C (4 mmHg)
Flash Point 166°C (330°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.519
Quantity 500 g
UN Number UN3082
Beilstein 1880877
Formula Weight 246.27
Percent Purity 97%
Chemical Name or Material Diallyl phthalate
Show More Show Less
Hazard Category H302-H317-H331-H340-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P311-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 205-016-3
RTECSNumber CZ4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only