accessibility menu, dialog, popup

UserName

IN

CAS RN 130723-54-5

IUPAC Name: 2-(3-iodophenyl)acetonitrile
Synonyms: 2-(3-iodophenyl)acetonitrile
Molecular Weight (g/mol): 243.05
Molecular Formula: C8H6IN
InChi Key: LVOKGAHCTHNWIL-UHFFFAOYSA-N
SMILES: IC1=CC=CC(CC#N)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)