missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenylacetonitrile, 97%

Catalog No. AAL1511806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1511806 5 g
AAL1511803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1511806 Supplier Thermo Scientific Chemicals Supplier No. L1511806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 130723-54-5 | C8H6IN | 243.05 g/mol

3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 130723-54-5
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.05
MDL Number MFCD00040890
InChI Key LVOKGAHCTHNWIL-UHFFFAOYSA-N
Synonym 3-iodophenylacetonitrile, 2-3-iodophenyl acetonitrile, 3-iodobenzyl cyanide, 3-iodophenyl acetonitrile, m-iodobenzyl cyanide, 3-iodophenyacetonitrile, pubchem12771, 3-iodo-1-phenylacetonitrile, acmc-1c0v0, benzeneacetonitrile, 3-iodo
PubChem CID 2759368
IUPAC Name 2-(3-iodophenyl)acetonitrile
SMILES IC1=CC=CC(CC#N)=C1

Specifications

Melting Point 33°C to 35°C
Boiling Point 138°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Beilstein 7350695
Sensitivity Light sensitive
Solubility Information Insoluble in water
Formula Weight 243.05
Percent Purity 97%
Chemical Name or Material 3-Iodophenylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only