accessibility menu, dialog, popup

UserName

CH3OOFFFFFFFFF

CAS RN 13038-26-1

IUPAC Name: methyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
Molecular Weight (g/mol): 278.07
Molecular Formula: C6H3F9O2
InChi Key: OSDPSOBLGQUCQX-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results