missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nonafluorovalerate 97.0+%, TCI America™

Catalog No. M19125G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19125G Supplier TCI America Supplier No. M19125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13038-26-1
Molecular Formula C6H3F9O2
Molecular Weight (g/mol) 278.074
MDL Number MFCD00443384
InChI Key OSDPSOBLGQUCQX-UHFFFAOYSA-N
Synonym Methyl Perfluorovalerate, Methyl Nonafluoropentanoate, Methyl Perfluoropentanoate, Nonafluorovaleric Acid Methyl Ester, Perfluorovaleric Acid Methyl Ester
PubChem CID 526458
IUPAC Name methyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
SMILES COC(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 3272
Formula Weight 278.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonafluorovalerate