accessibility menu, dialog, popup

UserName

NHNH2

CAS RN 13035-19-3

IUPAC Name: piperidin-4-amine
Synonyms: piperidin-4-amine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: BCIIMDOZSUCSEN-UHFFFAOYSA-N
SMILES: NC1CCNCC1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)