accessibility menu, dialog, popup

UserName

CAS RN 128073-16-5

IUPAC Name: ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Synonyms: ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Molecular Weight (g/mol): 253.61
Molecular Formula: C9H7ClF3NO2
InChi Key: COMQYNZHBCNPNW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Cl)C=C(C=N1)C(F)(F)F
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)