accessibility menu, dialog, popup

UserName

CAS RN 128-04-1

IUPAC Name: sodium (dimethylcarbamothioyl)sulfanide
Synonyms: dimethyldithiocarbamic acid, sodium salt
Molecular Weight (g/mol): 143.2
Molecular Formula: C3H6NNaS2
InChi Key: VMSRVIHUFHQIAL-UHFFFAOYSA-M
SMILES: [Na+].CN(C)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results