missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dimethyldithiocarbamate Dihydrate 98.0+%, TCI America™

Catalog No. D0716500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0716500G 500 g
D071625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0716500G Supplier TCI America Supplier No. D0716500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128-04-1
Molecular Formula C3H6NNaS2
Molecular Weight (g/mol) 143.198
MDL Number MFCD00150729
InChI Key VMSRVIHUFHQIAL-UHFFFAOYSA-M
Synonym Dimethyldithiocarbamic Acid Sodium Salt
PubChem CID 560256
IUPAC Name sodium;N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)[S-].[Na+]

Specifications

Color White
Quantity 500 g
UN Number 3077
Formula Weight 143.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dimethyldithiocarbamate Dihydrate
EINECSNumber (2)-1249&(2)-1833
RTECSNumber FD3500000
TSCA Yes