accessibility menu, dialog, popup

UserName

CH3H3CNCH3OClOOO(?)

CAS RN 1276539-32-2

IUPAC Name: 9-(2,6-dimethylphenyl)-10-methylacridin-10-ium perchlorate
Synonyms: 9-(2,6-dimethylphenyl)-10-methylacridin-10-ium perchlorate
Molecular Weight (g/mol): 397.86
Molecular Formula: C22H20ClNO4
InChi Key: AUILEGHJOMNNKY-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC1=CC=CC(C)=C1C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results