missing translation for 'onlineSavingsMsg'
Learn More

9-(2,6-Dimethylphenyl)-10-methylacridinium Perchlorate, TCI America™

Catalog No. D34291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34291G Supplier TCI America Supplier No. D34291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1276539-32-2
Molecular Formula C22H20ClNO4
Molecular Weight (g/mol) 397.855
InChI Key AUILEGHJOMNNKY-UHFFFAOYSA-M
PubChem CID 44629820
IUPAC Name 9-(2,6-dimethylphenyl)-10-methylacridin-10-ium;perchlorate
SMILES CC1=C(C(=CC=C1)C)C2=C3C=CC=CC3=[N+](C4=CC=CC=C42)C.[O-]Cl(=O)(=O)=O

Specifications

Color Yellow
Quantity 1 g
UN Number 1479
Formula Weight 397.86
Physical Form Crystalline Powder
Chemical Name or Material 9-(2,6-Dimethylphenyl)-10-methylacridinium Perchlorate