accessibility menu, dialog, popup

UserName

CH3OOFFClFF

CAS RN 127589-63-3

IUPAC Name: methyl 3-chloro-2,2,3,3-tetrafluoropropanoate
Synonyms: methyl 3-chloro-2,2,3,3-tetrafluoropropanoate
Molecular Weight (g/mol): 194.51
Molecular Formula: C4H3ClF4O2
InChi Key: YHMRYDXPCZSESQ-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results