missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Chloro-2,2,3,3-tetrafluoropropionate 97.0+%, TCI America™

Catalog No. M19025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19025G 5 g
M19021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19025G Supplier TCI America Supplier No. M19025G
Only null left

Chemical Identifiers

CAS 127589-63-3
Molecular Formula C4H3ClF4O2
Molecular Weight (g/mol) 194.51
MDL Number MFCD00155715
InChI Key YHMRYDXPCZSESQ-UHFFFAOYSA-N
Synonym 3-Chloro-2,2,3,3-tetrafluoropropionic Acid Methyl Ester
PubChem CID 2782496
IUPAC Name methyl 3-chloro-2,2,3,3-tetrafluoropropanoate
SMILES COC(=O)C(F)(F)C(F)(F)Cl

Specifications

Quantity 5 g
UN Number 2924
Formula Weight 194.51
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Chloro-2,2,3,3-tetrafluoropropionate