accessibility menu, dialog, popup

UserName

CAS RN 1274892-48-6

IUPAC Name: bis(3-(ethylamino)phenol); sulfuric acid
Synonyms: bis(3-(ethylamino)-phenol); sulfuric acid
Molecular Weight (g/mol): 372.44
Molecular Formula: C16H24N2O6S
InChi Key: CLHKUUQJWJTXSF-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CCNC1=CC(O)=CC=C1.CCNC1=CC(O)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results