missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethylamino)phenol Hemisulfate 98.0+%, TCI America™

Catalog No. E05345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05345G 5 g
E05341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05345G Supplier TCI America Supplier No. E05345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1274892-48-6
Molecular Formula C16H24N2O6S
Molecular Weight (g/mol) 372.436
MDL Number MFCD03093630
InChI Key CLHKUUQJWJTXSF-UHFFFAOYSA-N
PubChem CID 44630284
IUPAC Name 3-(ethylamino)phenol;sulfuric acid
SMILES CCNC1=CC(=CC=C1)O.CCNC1=CC(=CC=C1)O.OS(=O)(=O)O

Specifications

Color Gray-White
Quantity 5 g
Formula Weight 186.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Ethylamino)phenol Hemisulfate