CAS RN 127-66-2
IUPAC Name:
2-phenylbut-3-yn-2-ol
Synonyms:
2-phenylbut-3-yn-2-ol
Molecular Weight (g/mol):
146.19
Molecular Formula:
C10H10O
InChi Key:
KSLSOBUAIFEGLT-UHFFFAOYNA-N
SMILES:
CC(O)(C#C)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Phenyl-3-butyn-2-ol, 98%
2-Phenyl-3-butyn-2-ol, 98%
CAS: 127-66-2 Molecular Formula: C10H10O Molecular Weight (g/mol): 146.189 MDL Number: MFCD00004454 InChI Key: KSLSOBUAIFEGLT-UHFFFAOYSA-N Synonym: 2-phenyl-3-butyn-2-ol,methylphenylethynylcarbinol,3-hydroxy-3-phenyl-1-butyne,3-butyn-2-ol, 2-phenyl,3-phenyl-butin-1-ol-3,alpha-ethynyl-alpha-methylbenzyl alcohol,alpha-ethynyl-alpha-methylbenzenemethanol,benzenemethanol, .alpha.-ethynyl-.alpha.-methyl,3-phenyl-butin-1-ol-3 german,benzenemethanol, alpha-ethynyl-alpha-methyl PubChem CID: 92098 IUPAC Name: 2-phenylbut-3-yn-2-ol SMILES: CC(C#C)(C1=CC=CC=C1)O