accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 127-17-3

IUPAC Name: 2-oxopropanoic acid
Synonyms: pyruvic acid acid, pyruvic 2-oxopropionsaeure BTS pyruvic acid (substance) pyroracemate brenztraubensaeure ion(1-) 2-oxopropanoic acid α-oxopropionsaeure 2-oxopropanoate
Molecular Weight (g/mol): 88.06
Molecular Formula: C3H4O3
InChi Key: LCTONWCANYUPML-UHFFFAOYSA-N
SMILES: CC(=O)C(O)=O
Boiling Point 
  • (3)
  • (6)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (6)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (14)
Grade 
  • (4)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (6)
  • (6)
  • (2)
  • (7)
  • (3)
  • (6)
  • (4)
  • (2)
  • (2)
  • (6)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (14)
  • (4)
  • (6)