missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid 97.0+%, TCI America™

Catalog No. P0579500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0579500G 500 g
P057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0579500G Supplier TCI America Supplier No. P0579500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-17-3
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
MDL Number MFCD00002585
InChI Key LCTONWCANYUPML-UHFFFAOYSA-N
Synonym pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
PubChem CID 1060
ChEBI CHEBI:32816
IUPAC Name 2-oxopropanoic acid
SMILES CC(=O)C(O)=O

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 62°C
Quantity 500 g
UN Number 3265
Formula Weight 88.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyruvic Acid
EINECSNumber (2)-0959&(2)-1473
RTECSNumber UZ0829800
TSCA Yes
Recommended Storage Refrigerator