missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid, 95-100%, Spectrum™ Chemical

Catalog No. 18613006
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613006 500 g Amber Glass Bottle
18613003 5 g Amber Glass Bottle
18613000 100 g Amber Glass Bottle
18613004 5 kg Amber Glass Bottle
18613001 12 kg Amber Glass Bottle
18613005 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613006 Supplier Spectrum Chemical Mfg Cor Supplier No. PY110500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-17-3

Specifications

CAS Max % 1
Boiling Point 165°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 95 to 100%
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.