accessibility menu, dialog, popup

UserName

CH3HOCl

CAS RN 127-00-4

IUPAC Name: 1-chloropropan-2-ol
Synonyms: 1-chloro-2-propanol
Molecular Weight (g/mol): 94.54
Molecular Formula: C3H7ClO
InChi Key: YYTSGNJTASLUOY-UHFFFAOYNA-N
SMILES: CC(O)CCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)