missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-propanol (contains ca. 25% 2-Chloro-1-propanol) 70.0+%, TCI America™

Catalog No. C0269500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0269500ML 500 mL
C026925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0269500ML Supplier TCI America Supplier No. C0269500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-00-4
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00004530
InChI Key YYTSGNJTASLUOY-UHFFFAOYSA-N
Synonym 1-chloro-2-propanol, 2-propanol, 1-chloro, chloroisopropyl alcohol, propene chlorohydrin, 1-chloroisopropyl alcohol, sec-propylene chlorohydrin, 1-chloro-2-hydroxypropane, .alpha.-propylene chlorohydrin, ccris 4766, 3-chloro-2-propanol
PubChem CID 31370
ChEBI CHEBI:76260
IUPAC Name 1-chloropropan-2-ol
SMILES CC(CCl)O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 500 mL
UN Number 2611
Formula Weight 94.54
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-propanol (contains ca. 25% 2-Chloro-1-propanol)
EINECSNumber (2)-2002
RTECSNumber UA8942000
TSCA Yes