accessibility menu, dialog, popup

UserName

CAS RN 1268340-93-7

BNHFFF

CAS RN 1268340-93-7

IUPAC Name: trifluoro[(piperidin-1-ium-1-yl)methyl]boranuide
Synonyms: trifluoro(piperidin-1-ium-1-ylmethyl)boranuide
Molecular Weight (g/mol): 166.98
Molecular Formula: C6H13BF3N
InChi Key: PFMNVTAJBBIDDU-UHFFFAOYSA-O
SMILES: F[B-](F)(F)C[NH+]1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results