missing translation for 'onlineSavingsMsg'
Learn More

(Piperidinium-1-ylmethyl)trifluoroborate 96.0+%, TCI America™

Catalog No. P22871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P22871G Supplier TCI America Supplier No. P22871G
Only null left

Chemical Identifiers

CAS 1268340-93-7
Molecular Formula C6H13BF3N
Molecular Weight (g/mol) 166.982
MDL Number MFCD18071160
InChI Key PFMNVTAJBBIDDU-UHFFFAOYSA-O
Synonym piperidinium-1-ylmethyl trifluoroborate, piperidinium-1-ylmethyltrifluoroboronanion, trifluoro piperidin-1-ium-1-ylmethyl boranuide, trifluoro piperidin-1-ium-1-yl methyl borate 1-, piperidinium-1-ylmethyl trifluoroborate, internal salt
PubChem CID 53243642
IUPAC Name trifluoro(piperidin-1-ium-1-ylmethyl)boranuide
SMILES [B-](C[NH+]1CCCCC1)(F)(F)F

Specifications

Melting Point 150°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 166.98
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (Piperidinium-1-ylmethyl)trifluoroborate