accessibility menu, dialog, popup

UserName

CAS RN 126198-74-1

H3CCH3OOOO

CAS RN 126198-74-1

IUPAC Name: 1-benzyl 2-(7-methyloctyl) benzene-1,2-dicarboxylate
Synonyms: 1-benzyl 2-(7-methyloctyl) phthalate
Molecular Weight (g/mol): 382.5
Molecular Formula: C24H30O4
InChi Key: IZTZLYGDXPHVQF-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results