missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Isononyl Phthalate (mixture of branched chain isomers), TCI America™
Supplier: TCI America B271525G
Specifications
| Benzyl Isononyl Phthalate (mixture of branched chain isomers) | |
| Yellow | |
| 25 g | |
| IZTZLYGDXPHVQF-UHFFFAOYSA-N | |
| 2-O-benzyl 1-O-(7-methyloctyl) benzene-1,2-dicarboxylate | |
| 57347326 | |
| Liquid |
| 126198-74-1 | |
| C24H30O4 | |
| Phthalic Acid Benzyl Isononyl Ester | |
| CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCC2=CC=CC=C2 | |
| 382.5 | |
| 382.50 |
Chemical Identifiers
| 126198-74-1 | |
| 382.5 | |
| Phthalic Acid Benzyl Isononyl Ester | |
| 2-O-benzyl 1-O-(7-methyloctyl) benzene-1,2-dicarboxylate |
| C24H30O4 | |
| IZTZLYGDXPHVQF-UHFFFAOYSA-N | |
| 57347326 | |
| CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCC2=CC=CC=C2 |
Safety and Handling
EINECSNumber : (3)-1312
TSCA : No