accessibility menu, dialog, popup

UserName

CH3CH3H2NHO

CAS RN 124-68-5

IUPAC Name: 2-amino-2-methylpropan-1-ol
Synonyms: 2-amino-2-methyl-1-propanol aminomethyl propanol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: CBTVGIZVANVGBH-UHFFFAOYSA-N
SMILES: CC(C)(N)CO
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)